Tashiro’s indicator solution i etanol, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
:
MDL Number:MFCD00148563
Boiling Pt:78 °C (1,013 hPa)
Density:0,79 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Omgivande
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Change range (pH 5-6):complying