Kongoröd
Catalog # EHERC12965400
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Förp.:0,1 g
- Seq:3020
Specifications
About this item
:
:696.67 g/mol
Merck Index:12,02562
Melting Pt:> 360 °C
EINECS:209-358-4
:GHS08
Storage Temperature:Omgivande
CAS Number:573-58-0
Formula:C₃₂H₂₂N₆Na₂O₆S₂
MDL Number:MFCD00004028