Waistcoats, cold protection, Bernex
Supplier: SIOEN
Certificates
About this item
Body warming waistcoats made of 100% polyester ripstop with a PVC coating and stitched seams. The garments feature two patched chest pockets with gusset, two patched pockets, one inside pocket, phone pocket, pen pocket and one loop for badge.
- Extended back and straight collar
- Zip closure under flap with press studs
- Drawstring in hem
- Fixed quilted lining inside
Specifications
- Catalog No:
- 113-6074
- 113-6073
- SIOE087A/B81/3XL
- SIOE087A/B81/2XL
- 113-6072
- 113-6071
- SIOE087A/B81/1XL
- 113-6067
- SIOE087A/B81/L
- SIOE087A/B81/S
- 113-6068
- SIOE087A/B81/M
- Modell:
- Bernex
- Bernex
- Bernex
- Bernex
- Bernex
- Bernex
- Bernex
- Bernex
- Bernex
- Bernex
- Bernex
- Bernex
- Outfit applikationer:
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- General Purpose
- Risk area:
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Cold
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating
- 100% polyester ripstop with a PVC coating