Tray
155×95×24 mm
Katalog # TSLA73034
Leverantör: TECHNICAL QUARTZ SOLUTIONS
Gå till:
Produktdetaljer och dokument
Trays made of quartz.
- Temperature resistance up to +1050 °C
- Highly resistant to chemicals
Specifikationer
- B×D×H:155×95×24 mm