N,N-Dimetylacetamid, SupraSolv® for headspace gas chromatography, Supelco®
Catalog # 1.00399.1000
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:1 L
- Seq:5037
Specifications
About this item
:
:87.12 g/mol
Merck Index:13,03254
Boiling Pt:166,1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0,937 g/cm³ (20 °C)
Flash Pt:70 °C
EINECS:204-826-4
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS08
:GHS07
Storage Temperature:Omgivande
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686