Microcentrifuge tubes, 1,5 ml
Catalog # LPIT111540
Supplier: LP ITALIANA
Specifications
- Typ:with attached snap cap, Eppendorf type
- Version:Graduated, conical bottom, non sterile
- Kapacitet:1,5 ml
- Färg:Ofärgad
- Förp.:10.000
Specifications
About this item
PP, translucent.
- Excellent chemical resistance
- Excellent temperature resistance
- Graduated.
With flat cap.