Shelf for baskets
Katalog # METOCC9744A
Leverantör: Metro International
Specifikationer
- Beskrivning:Basket shelf, 914×457 mm
- Förp.:1
Specifikationer
Om denna produkt
Heavy duty construction baskets are 89 mm deep and can hold upto 182 kg of weight. Ideal for containing and displaying smaller items.