Shelf, heavy duty dunnage, Super Erecta Shelf®
Katalog # METO1824DRC
Leverantör: Metro International
Specifikationer
- Beskrivning:Shelves, heavy duty dunnage, Super Erecta Shelf®, DR serie
- Material:Förkromad
- B×D×H:610×457×1880 mm
- Förp.:1
- Vikt:6,3 kg
Specifikationer
Om denna produkt
The heavy-duty dunnage shelves enhance the weight bearing capacity the system.
- Available with removable wire mat for easy lifting and to clean
- Support frames: 25 mm square tubing lifts off for easy cleaning