Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Mortlar och pistiller
Pistiller
VWR®, Pestles
VWR®, Pestles
VWR®, Pestles
Leverantör:
VWR Collection
Certifikat
Expandera bilden
VWR®, Pestles
Leverantör:
VWR Collection
Om denna produkt
Porcelain.
Grinding surface unglazed
Glazed handle
Certifications:
Produced according to DIN 12906.
Relaterad information
Dokumentation
Bilder 360
410-0122
CoC VWR Pestles
Dokumentation