Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Kranar
Standard valves
Standard valves
Standard valves
Leverantör:
COWIE
Certifikat
Expandera bilden
Standard valves
Leverantör:
COWIE
Om denna produkt
Standard valves made of PTFE.
Maximum Pressure 1 bar at 100 °C and 5 mm Hg
Valves may be sterlised at 135 °C
Very good chemical resistance