Centrifuge tubes, conical bottom
Catalog # WITG4.945.005
Leverantör: witeg Labortechnik
Specifikationer
- Stängningsstorlek:-
- Ø×L:40×115 mm
- Kapacitet:85 ml
- Förp.:1
Specifikationer
Om denna produkt
DURAN®, borosilicate glass.
- Heavy wall
- Ungraduated
- Very good chemical resistance
- Very good temperature resistance
Conical bottom, without rim