Orders LinkContactUsLinkComponent
Automatic burettes, Pellet pattern, Class AS
Automatic burettes, Pellet pattern, Class AS
Supplier:   witeg Labortechnik
Automatic burettes, Pellet pattern, Class AS
Supplier:  witeg Labortechnik
Certificates
Automatic burettes, Pellet pattern, Class AS
Automatic burettes, Pellet pattern, Class AS
2 Options Available Below
Product Details & Documents
Automatic burettes, Pellet pattern, class AS, conformity certified, made of DURAN.
  • Main point ring-graduation
  • Without stopcock between burette and reservoir
  • Lateral needle valve stopcock, with PTFE-needle valve
  • With overtwist device
  • With 2000 ml reservoir bottle, without aspirator bulb
Product Family Options
Product Information
  • Beskrivning
    Kapacitet
    Indelning
    Tolerans
    Availability
    Price
  • Brunfärgat glas
    50 ml
    0,1 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 10 350,00 kr
    10 350,00 kr
    1 * 2 Items
       10 350,00 kr
  • Med blå Schellbach-remsa
    50 ml
    0,1 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 2 Items
    1 * 2 Items - 6 975,00 kr
    6 975,00 kr
    1 * 2 Items
       6 975,00 kr