Orders LinkContactUsLinkComponent
Separating funnels, Squibb
Separating funnels, Squibb
1000 ml, 29/32 NS
Katalog # WITG2.658.001
Leverantör:   witeg Labortechnik
Separating funnels, Squibb
Katalog # WITG2.658.001
Leverantör:  witeg Labortechnik
Separating funnels, Squibb
Separating funnels, Squibb
1000 ml, 29/32 NS
Price
1 400,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Separating funnels, Squibb, made of borosilicate glass, stopcock with PTFE key. Complete wth PE stopper.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Specifikationer
  • NS-hylsa:
    29/32 NS
  • Kapacitet:
    1000 ml