Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Pipetter
Vollpipetter
Bulb pipette, FORTUNA®, class AS
Bulb pipette, FORTUNA®, class AS
Bulb pipette, FORTUNA®, class AS
Leverantör:
Poulten & Graf
Certifikat
Expandera bilden
Bulb pipette, FORTUNA®, class AS
Leverantör:
Poulten & Graf
Om denna produkt
Clear glass.
Two marks
Blue or amber graduation
Conformity approved, with batch certificate