Burettes, straight stopcock, Schellbach, FORTUNA®
Catalog # 612-0564
Supplier: Poulten & Graf
Specifications
- Kapacitet:10 ml
- Indelning:0,02 ml
- Förp.:1
Specifications
About this item
Borosilicate glass 3.3, class AS.
- Calibrated to deliver 'Ex'
- Schellbach stripe, blue enamel graduations
*Short form
Certifications: EN ISO 385