Splash heads, bent, acc. to Stutzer
Catalog # WITG1.425.005
Supplier: witeg Labortechnik
Specifications
- Kärna:24/29 NS
- Förp.:1
Specifications
About this item
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Two standard ground cones, inclined at an angle of 75°. Placed directly on the distillation flask to intercept droplets.