Splash heads, acc. to Stutzer, straight
Katalog # WITG1.420.004
Leverantör: witeg Labortechnik
Specifikationer
- Kärna:29/32 NS
- NS-hylsa:29/32 NS
- Förp.:1
Specifikationer
Om denna produkt
Borosilicate glass 3,3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Placed directly on the distillation flask to intercept droplets, with two standard ground joints.