Distilling heads, Claisen type
Katalog # WITG1.020.004
Leverantör: witeg Labortechnik
Specifikationer
- Kärna till kondensator:19/26 NS
- Propp till kolv:24/29 NS
- Förp.:1
Specifikationer
Om denna produkt
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Acc. to DIN 12594 and in addition to standard, with two parallel NS sockets and two NS cones (descending angle: 75°), for distillation thermometers of 55 mm built-in length.