ContactUsLinkComponent
Transport container, Crioplast®
Transport container, Crioplast®
Catalog # LPIT100015_U
Supplier:  LP ITALIANA
undefined
Transport container, Crioplast®
Catalog # LPIT100015_U
Supplier:  LP ITALIANA
Specifications
  • Beskrivning:
    Container for transporting blood, suitable for 16×100 mm tubes
  • B×D×H:
    181×257×274 mm
  • Förp.:
    4
Specifications
About this item
Plastic, white, with blue inserts.
Crioplast® refrigerated container for maintaining and transporting blood at original conditions.