Conical sleeves for vacuum filtration, Sterilin®
Katalog # 511-0098
Leverantör: STERILIN
Specifikationer
- Beskrivning:Set of 6 cones, assorted
- Förp.:1 SET
Specifikationer
Om denna produkt
Red natural rubber cones for use with filtering flasks.
- Set of six pieces
- Set includes each one of 7/15 mm, 10/22 mm, 15/28 mm, 16/36 mm, 22/42 mm and 26/52 mm (inner diamater at low end/inner diameter at top end)