Olje rött O, Certistain® för mikroskopi, Sigma-Aldrich®
Catalog # 1.05230.9999
Supplier: Merck
CAS Number:
Specifications
- Förp.:1
- Seq:0001
Specifications
Specification Test Results
- Dye content (spectrophotometrically):≥ 80 %
- Identity (UV/VIS-Spectrum):passes test
- Absorption maximum λ max (chloroform):528 - 534 nm
- Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; chloroform):725 - 910
- TLC-Test:passes test
- Loss on drying (110 °C):≤ 8 %
- Suitability for microscopy:passes test
Specification Test Results
About this item
:
:408.51 g/mol
:M-Clarity™ quality level = MQ300
Storage Temperature:Omgivande
EINECS:215-295-3
:1
CAS Number:1320-06-5
Formula:C₂₆H₂₄N₄O
MDL Number:MFCD00003898