Rotor for Cryofuge™ 6000i/8500i series
Catalog # 521-1384
Supplier: Thermo Scientific
Specifications
- Beskrivning:Vinskyddad rotor med lock, 4 020/5 050 min⁻¹
- Seq:01
- Förp.:1
Specifications
About this item
Windshielded rotor for the large capacity centrifuges Cryofuge 6000i and 8500i.
- Swinging bucket
- Rotor body made of stainless steel; bucket made of aluminium
- Comes with a lid but without buckets