Hydrophilic interaction chromatography columns, Asahipak NH2P preparative type
: Resonac
Asahipak NH2P are polyvinyl alcohol-based packed columns ideal for saccharides analysis.
- Excellent chemical stability and resistance to degradation over time
- Washable with alkaline solutions
- Suitable for evaporative light scattering detector, corona-charged aerosol detector and LC-MS
- Catalog No:
- 554-5307
- 554-5308
- 554-5306
- Benämning:
- Asahipak NH2P-90 20F
- Asahipak NH2P-130G 7B
- Asahipak NH2P-50 10E
- Max. tryck:
- 10 MPa (100 bar)
- 15 MPa (150 bar)
- pH område:
- 2 - 13
- 2 - 13
- 2 - 13
- Temperature limit:
- 4 - 50 °C
- 4 - 50 °C
- 4 - 50 °C
- USP code:
- L82
- L82
- L82