Transport swabs (Sterilin)
Leverantör: STERILIN
Certifikat
Om denna produkt
These swabs formulation includes antibiotics to inhibit bacterial and fungal flora in samples.
- Room temperature stability via a cryoprotectant
- Glass beads help with the release and dispersion of the sample
- Free standing skirted tube with conical base: Simply vortex and inoculate
Designed for the collection, transport, maintenance and frozen storage of viruses, Chlamydia, mycoplasma and ureaplasma in cell culture, rapid antigen detection, DFA, EIA, PCR and nucleic acid amplification assays.
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904