Beaker tongs, stainless steel
Catalog # NICK7005
Supplier: NICKEL ELECTRO
Specifications
- Längd:356 mm
- Förp.:1
Specifications
About this item
These stainless steel beaker tongs feature silicone covered jaws.
- Silicone covered sleeves reduce risk breakage when handling beakers
- Operating temperature: −60...+180˚C
- Hold beakers from 100 to 1000 ml