VWR® Ready Ladder™ DNA Ladder, 50 bp
Katalog # N746-100RXN
Leverantör: VWR Chemicals
CAS-nummer:
Specifikationer
- Förp.:1 KIT
- Beskrivning:50 bp Ready Ladder™, pre-mixed with loading dye, sufficient for 100 lanes
Specifikationer
Om denna produkt
Ladder with 10 fragments ranging from 50 to 500 bp for easy band identification. Ready to use formula pre-mixed with loading dye.