HPLC conventional columns, Brownlee™ Spheri-5®
Leverantör: PerkinElmer
Certifikat
Om denna produkt
Small pore silica-based sorbent used for separation of small molecules. Non-functional and polyfunctional C8 and C18 are available.
Small pore silica-based sorbent used for separation of small molecules. Non-functional and polyfunctional C8 and C18 are available.