Magnetic stirring bar, set
Catalog # HEID509-58300-00
Supplier: Heidolph Instruments GmbH & Co.KG
Specifications
- Typ:Set of cross shape, elliptical and oval megnetic stirrer bars
- Förp.:10
Specifications
About this item
Set of different shaped PTFE covered magentic stirring bars.
- Very good chemical resistance
- Temperature resistant, suitable for high and low temperature applications
Set includes 2× each of stirrers in a plastic box, stirrers have the following shapes and dimensions, L (mm): Cross shape 10 and 16,5 mm, elliptical shape 25 mm, oval 40 and 50 mm.