Magnetic stirring bars set, cylindrical
Catalog # HEID509-56000-00
Supplier: Heidolph Instruments GmbH & Co.KG
Specifications
- Typ:Set of 3 cylindrical magnetic stirring bars
- Förp.:1 SET
Specifications
About this item
Set of PTFE covered magentic stirring bars.
- Very good chemical resistance
- Temperature resistant, suitable for high and low temperature applications
Set includes 1× each of stirrers with the following dimensions, L (mm): 25, 40 and 50 mm