N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-8, SB-8), Millipore®
Catalog # 693019-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:5 g
- Seq:3173
Specifications
About this item
:
:GHS07
Storage Temperature:Omgivande
CAS Number:15178-76-4
Formula:C₁₃H₂₉NO₃S
MDL Number:MFCD00036912