Earmuffs for welding helmets, Peltor™ H505B
Catalog # MMMAH505B
Leverantör: 3M
Specifikationer
- Uppställning höjd:26 dB
- Färg:Svart
- Beskrivning:Peltor™ H505B welding helmet earmuffs with neck strap, suitable for use with a welding helmet
- Förp.:1
Specifikationer
Om denna produkt
These welding helmet earmuffs with neck strap have been designed for use with welding shields, particularly the 3M™ Speedglass™ 9100.
- Soft earcups filled with fluid for enhanced comfort
- The design of the neckband improves compatibility with other personal protective equipment
- Generous space underneath the earmuffs helps to reduce build-up of heat and sweat
- Soft, wide earcups guarantee optimum seal and low pressure for outstanding comfort
- Cushions and inserts are easy to replace and to keep clean and hygienic
The earmuffs provide moderate insulation, which is sufficient for most welding applications.
SNR: 26 dB
Certifications: EN 352-1:2002