Flame-retardant fleece jacket, Dampremy 7771
: SIOEN
Outdoor jacket made of double sided, flame retardant polyester fleece. With fixed flame retardant cotton lining on the inside. Can be zipped into different jackets.
- With anti-pilling treatment and stitched seams
- Straight collar, hidden zip closure
- Two inset pockets with zip closure
- Round inset sleeves, elasticated cuffs
- Elastic drawstring in hem
Weight: 275 g/m²
: EN 533 (index 3), EN 14058 (1XXXX)
- Catalog No:
- SIOE7771/B75/3XL
- SIOE7771/B75/2XL
- SIOE7771/B75/1XL
- SIOE7771/B75/L
- SIOE7771/B75/M
- SIOE7771/B75/S
- Modell:
- 7771 Dampremy
- 7771 Dampremy
- 7771 Dampremy
- 7771 Dampremy
- 7771 Dampremy
- 7771 Dampremy
- Outfit applikationer:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- Flame retardant cotton
- Flame retardant cotton
- Flame retardant cotton
- Flame retardant cotton
- Flame retardant cotton
- Flame retardant cotton
- Vikt:
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²