Flame retardant rain jacket, Hasnon 3085
Leverantör: SIOEN
Certifikat
Om denna produkt
Multifunctional jacket with taped seams, made of Siopor® FR AST consisting of 100% polyester fabric with 100% FR PU coating + AST. The fixed cotton lining is flame-retardant. The fabric is water repellent on the outside and features a moisture attracting coating on the inside. Different linings or fleeces can be zipped in.
- Highly breathable, comfortable, supple, high tear resistance
- Straight collar with foldaway hood
- Zip closure under flap with touch & close fastening
- Two inset chest pockets with zip closure, two patched pockets, phone pocket, Napoleon pocket, two loops for badges
- Round inset sleeves, narrowing by touch and close fastening
- Elasticated waist in the back + drawstring in the hem
Weight: 250 g/m²
Certifications: EN 13034 (Type PB/6), EN 531 (A/B2/C1), EN 14116, EN 1149-5, EN 343 (3-3)
Specifikationer
- Catalog No:
- 113-7087
- 113-7086
- 113-7089
- 113-7090
- 113-7091
- 113-7088
- Modell:
- 3085 Hasnon Siopor
- 3085 Hasnon Siopor
- 3085 Hasnon Siopor
- 3085 Hasnon Siopor
- 3085 Hasnon Siopor
- 3085 Hasnon Siopor
- Outfit applikationer:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Rain (waterproof), Flame-retardant
- Rain (waterproof), Flame-retardant
- Rain (waterproof), Flame-retardant
- Rain (waterproof), Flame-retardant
- Rain (waterproof), Flame-retardant
- Rain (waterproof), Flame-retardant
- Kön:
- För män
- För män
- För män
- För män
- För män
- För män
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 100% polyester with 100% FR PU coating + AST
- 100% polyester with 100% FR PU coating + AST
- 100% polyester with 100% FR PU coating + AST
- 100% polyester with 100% FR PU coating + AST
- 100% polyester with 100% FR PU coating + AST
- 100% polyester with 100% FR PU coating + AST
- Vikt:
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²