High visibility waistcoat, Elba 9044
Supplier: SIOEN
Certificates
About this item
Highly visible waistcoat made of mesh polyester in standard quality.
- V-neck
- Closure with touch and close fastening
- Stitched seams
Back length: 70 cm
Weight: 110 g/m²
Certifications: EN 471 (2-2)
Specifications
- Catalog No:
- 113-07175
- 113-07174
- 113-07165
- 113-07164
- 113-5986
- 113-07168
- 113-5987
- 113-07170
- 113-07169
- 113-07173
- 113-07166
- 113-07171
- Modell:
- 9044
- 9044
- 9044
- 9044
- 9044
- 9044
- 9044
- 9044
- 9044
- 9044
- 9044
- 9044
- Outfit applikationer:
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Industrial
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Stängningsstorlek:
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Tyg:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Vikt:
- 110 g/m²
- 110 g/m²
- 110 g/m²
- 110 g/m²
- 110 g/m²
- 110 g/m²
- 110 g/m²
- 110 g/m²
- 110 g/m²
- 110 g/m²
- 110 g/m²
- 110 g/m²