Chemical resistant overalls, KLEENGUARD* A71
Leverantör: KIMBERLY CLARK
Certifikat
Om denna produkt
These disposable overalls made from a laminate of barrier film and a non woven fabric provide protection against high pressure aqueous jets, inorganic acids and bases and a range of organic chemicals. The non woven fabric is anti-static and silicone-free which is important when working with paint or sensitive surfaces.
- Low lint and silicone-free for critical areas
- Stitched and over-taped seams to provide high strength and barrier properties to withstand high pressure aqueous jets
- With hood and zip closure with flap
Colour: Yellow
Certifications: Type 3/4/5, EN 1149-1, EN 14126, EN 1073-2, Cat. III
Specifikationer
- Catalog No:
- 113-6394
- 113-6391
- 113-6393
- 113-6390
- 113-6392
- Modell:
- A71
- A71
- A71
- A71
- A71
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Antistatisk:
- +
- +
- +
- +
- +
- Outfit applikationer:
- Chemical protection, Paint and lacquer, Laboratory, Waste water, Industrial
- Chemical protection, Paint and lacquer, Laboratory, Waste water, Industrial
- Chemical protection, Paint and lacquer, Laboratory, Waste water, Industrial
- Chemical protection, Paint and lacquer, Laboratory, Waste water, Industrial
- Chemical protection, Paint and lacquer, Laboratory, Waste water, Industrial