Mini Tubes, SP Bel-Art
Katalog # 391-9030
Leverantör: Bel-Art Products, a Part of SP
Specifikationer
- Kapacitet:1 ml
- Beskrivning:400×12 tube strips
- Förp.:4.800
Specifikationer
Om denna produkt
Made of virgin, medical grade polypropylene.
- Autoclavable
- Frozen to −20 °C
- Very good chemical resistance
Mini-Tubes offer a convenient and economical means of performing laboratory procedures using multi-channel pipettors of four, eight or twelve channels. Features an attachment bridge between tubes, forming a Mini-Tube strip; can be easily separated into single tubes by cutting apart.