Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Enzymer
Chicken lysozyme (Muramidase) (from egg white)
Chicken lysozyme (Muramidase) (from egg white)
:
Merck Millipore (Calbiochem)
Chicken lysozyme (Muramidase) (from egg white)
:
Merck Millipore (Calbiochem)
CAS-nr:
12650-88-3
Källa:
Egg white
Species:
Chicken
Storage conditions:
Freezer
Enzymnamn:
Lysozyme (Muramidase)