VWR® Proteomics Grade, Tris-glycine buffer
Catalog # M115-40L
Supplier: VWR Chemicals
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle for solids
- Förp.:40 L
- Beskrivning:Tris-glycine buffer, prepares 40 L of 1X Tris-glycine solution
Specifications
About this item
A pre-blended Tris-glycine powder for preparation of Laemmli SDS-PAGE running buffer. The 40 litre pack size prepares 40 litres of 1X Tris-glycine solution (0,025 M Tris base and 0,192 M glycine). Each Ready-Pack™ prepares 1 litre of a 10X concentrate.