Transport trolleys for baskets, EURO-STANDARD
Katalog # BOCH17333
Leverantör: BOCHEM
Specifikationer
- Beskrivning:Transport cart
- B×D×H:610×465×1360 mm
- Förp.:1
Specifikationer
Om denna produkt
Transport cart made of stainless steel used for transport baskets.
- Five hanging ties
- Two pushing handles
- Two guide and fixed rolls, two with lock
- Enclosure of rolls, zincked
- Excess length of enclosure