Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bägare
Butter melting beakers
Butter melting beakers
Butter melting beakers
Catalog # 614-0198
Supplier:
GERBER FUNKE
Supplier #:
5401
Expand image
Butter melting beakers
Catalog # 614-0198
Supplier:
GERBER FUNKE
Supplier #:
5401
Log in
for availability
Ø utv.
Select an Option
×
64 mm
Höjd
Select an Option
×
66 mm
Förp.
Select an Option
×
1
About this item
Product Details
Aluminium.
For the determination of water content in butter.
Product Details