ContactUsLinkComponent
Draining racks, Poxygrid®, SP Bel-Art
  149-0197
undefined
Draining racks, Poxygrid®, SP Bel-Art
  149-0197
  • Beskrivning:
    Avrinningsställ, med 14 hårnålsöglor och 12 raka stift
  • B×D×H:
    430×170×480 mm
  • Förp.:
    1
Racks for drying test tubes, beakers, flasks, and other labware. The stand has rubber tipped feet for non-skid use on a tabletop, or wire loops on the back for wall mounting.
  • Three rows of epoxy-coated hairpin-loop supports (14 in all) hold labware with 18 mm int.Ø necks or larger
  • Top row has 12 straight pins 102 mm long to accommodate smaller items
  • Polystyrene tray catches water as the labware drains