Rack for cuvettes, SP Bel-Art
Katalog # BELAF185150000
Leverantör: Bel-Art Products, a Part of SP
Specifikationer
- För:20 (2×10) cuvettes
- B×D×H:210×70×35 mm
- Förp.:1
Specifikationer
Om denna produkt
Made of white PP. Rack holds 10 mm light path spectrophotometer cuvettes in two tiers with numbered wells.
- Autoclavable
- Very good chemical resiatnce
- Easy to clean
The wells hold standard cuvettes with a 12,5 mm cross-section. A sloped top makes the second tier more accessible.