Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Värme- och fläktvärmeskåp
Rörugnar
Accessories for rotary tube furnaces
Accessories for rotary tube furnaces
Accessories for rotary tube furnaces
Catalog # NABE691404552
Supplier:
NABERTHERM
Expand image
Accessories for rotary tube furnaces
Catalog # NABE691404552
Supplier:
NABERTHERM
Specifications
Beskrivning:
Quartz glass tube with pimple, 106×100×1540 mm, for Tmax. 1100 °C and 1300 °C models
Förp.:
1
För:
RSR 120 - 500
Specifications
About this item
Product Details
Product Details