Tray, for Petri dish incubation, SP Bel-Art
Catalog # BELAF189830000
Supplier: Bel-Art Products, a Part of SP
Specifications
- B×D×H:349×137×22 mm
- Förp.:1
Specifications
About this item
Tray made of white epoxy-coated steel.
- Excellent aid for keeping cell cultures organised
- With 29 holes of 19 mm diameter in bottom allow air passage
- Holds 100 mm petri dishes securely on an incubator rack