Laboratory glass cleaner, deconex® 21 SOLID
Catalog # 148-5191
Supplier: Borer Chemie
Specifications
- Beskrivning:deconex 21 SOLID
- Förp.:10 kg
Specifications
About this item
Mildly alkaline cleaning powder for automated cleaning of laboratory glassware und reusable laboratory material.
- Rinsable without residues and guarantees therefore consistent and error-free results
- With a broad cleaning spectrum and excellent results even at low concentrations
- High cleaning performance due to excellent dispersing capabilities
- Good material compatibility, especially recommended for alkali and corrosion sensitive items, also suitable for aluminium
- EDTA-, NTA-, chlorine- and surfactant-free, no release of surfactants into waste water
- Rinsing with an acidic deconex® cleaner is recommended as the completion of the cleaning process