Cap for BOD bottle, WHEATON®
Katalog # WHEA227723
Leverantör: DWK Life Sciences
Specifikationer
- Beskrivning:PP-propp, med skuminlägg
- Förp.:50
Specifikationer
Om denna produkt
This polyethylene cap, complete with a closed cell foam insert, snaps firmly over the flared mouth of Wheaton BOD bottles.
- Polyethylene cap prevents evaporation
- Includes foam insert
This closure prevents the evaporation of the water seal during the five-day BOD incubation period. The foam insert fits into the cap and exerts pressure on the bottle stopper to hold it securely in place.