Counting chamber, McMaster
Catalog # 630-1507
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Typ:McMaster
- Beskrivning:Cell counter
- Förp.:1
Specifications
About this item
Counting chamber with cover glass and two counting areas.
Specially designed for blood tests and urine analysis.