VWR®, Pipettspets, robot, till Beckman® Biomek FX/NX
Katalog # 736-0611
Leverantör: VWR Collection
Specifikationer
- CE-IVD:+
- Förp.:10×96
- Steril:-
- Volym:100 µl
- Seq:03
- Environmentally Preferable:Low Manufacturing Impact,Sustainable Packaging
- Tip style:Standard
- Filter:-
Specifikationer
Om denna produkt
Genomskinlig PP, autoklaverbara.
- RNase/DNase, DNA and endotoxin-free
- Sustainable design based on virgin resins