Duk, oljeabsorberande, T-151
121-1009
: 3M
- Kapacitet:140 L
- Färg:Vit
- L×B:430×480 mm
- Förp.:100/box, 2 boxes/case
Ideal for absorbing oil from areas where water may also be present.
Absorbent sheets are conveniently sized and can be quickly spread on and in the path of a small spill.
Each box contains 100 pads (430 mm x 480 mm) and sorbs 140 liter in total.