ContactUsLinkComponent
Absorbent powder, Chemizorb® Alkalis
Absorbent powder, Chemizorb® Alkalis
Catalog # 1.01596.1000
Supplier:  Merck
undefined
Absorbent powder, Chemizorb® Alkalis
Catalog # 1.01596.1000
Supplier:  Merck
Specifications
  • Beskrivning:
    Absorbent powder, Chemizorb® OHˉ
  • Förp.:
    Bottle
Specifications
About this item
Chemizorb Alkalis is a powder mixture that consists of a mineral co-polymer as the absorbent, an acidic salt as the neutraliser, and a pH indicator.
  • Easy to use
  • pH indicator makes it possible to monitor the neutralisation process
  • 100 to 120 g of Chemizorb OHˉ required to absorb and neutralise 100 ml of alkalis
Upon contact with the alkalis the absorbent first turns blue in colour. After neutralisation is complete, the blue colour becomes paler.